Fast Blue RR, Sigma-Aldrich®
Supplier: SIGMA-ALDRICH MICROSCOPY
Warning
201545-5GEA 63.1 EUR
201545-5G
Fast Blue RR, Sigma-Aldrich®
Fast Blue RR
Fast Blue RR has been used to detect alkaline phosphatase activity.
Formula: C₁₅H₁₆N₂O₃
MW: 272,30 g/mol
Melting Pt: 165-168 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008369
CAS Number: 6268-05-9