AriaMx real-time PCR system
Supplier: AGILENT
5190-7702 5190-7708 5190-9370 600906 600907 401425 401427 401490 401491 401492 401493 401494 G5380AA G5381AA G8830-67001 G8830-67002 G8830-67003 G8830-67004 G8830-67005 G8830-67006 401428
AGLS5190-7702EA 614 EUR
AGLS5190-7702 AGLS5190-7708 AGLS5190-9370 AGLS600906 AGLS600907 AGLS401425 AGLS401427 AGLS401490 AGLS401491 AGLS401492 AGLS401493 AGLS401494 732-3776 732-3777 732-3778 732-3779 732-3780 732-3781 732-3782 732-3783 AGLS401428
AriaMx real-time PCR system
PCR Cyclers
The AriaMx real-time PCR system is a fully integrated quantitative PCR amplification, detection and data analysis system.
  • Gene expression analysis
  • New genotyping/HRM capability, mRNA quantification
  • NGS quantification: Library preparation, result validation, nucleic acid monitoring and rare allele detection
  • Can use self-installable optical cartridges
  • Operates at a Cq uniformity standard deviation of <0,20 at fast cycling (5 s at 95 °C/10 s at 60 °C)
  • Data acquisition time of <3 s/plate
  • Dynamic range of nine orders of magnitude
  • Operates at a maximum altitude of 2000 m and in 20 to 80% relative humidity
The system design combines a state-of-the-art thermal cycler, an advanced optical system with an LED excitation source, and complete data analysis software.
This instrument can hold up to six optics modules, and the scanning optics design delivers optimal separation between the dyes and between samples also providing a 'closed tube' PCR detection format that can be used with a variety of fluorescence detection chemistries including SYBR® Green and EvaGreen dyes, as well as fluorogenic probe systems including TaqMan probes.
The AriaMx amplifies the productivity with its unique modular and flexible design, intuitive touch screen interface, advanced, easy to use reporting and 120+ attributes monitored through the built-in on-board diagnostics to help pinpoint assay or instrument issues as they arise.
Informace pro objednávání: Software upgrade statements available online, no charge. Optional for purchase warranty length of five years.
Includes configurable optical modules (up to six), a touch screen controller, Experimental Import, LIMS connectivity, 96 wells, 5 GB of protocol storage, on-board instrument diagnostics, remote monitoring capabilities, one year standard warranty.
Upozornení: For research use only. Not for use in diagnostic procedures.