Platinum nominally 50%, Ruthenium nominally 25% on high surface area advanced carbon support
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
47371.03 47371.06
47371.03EA 278 EUR
47371.03 47371.06
Platinum nominally 50%, Ruthenium nominally 25% on high surface area advanced carbon support
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Merck Index: 12,07689