8-Hydroxyquinoline-β-D-glucopyranoside
Supplier: Apollo Scientific
Warning
BI4356-100MG BI4356-250MG BI4356-50MG
APOSBI4356-100MGEA 174 EUR
APOSBI4356-100MG APOSBI4356-250MG APOSBI4356-50MG
8-Hydroxyquinoline-β-D-glucopyranoside
8-Hydroxyquinoline-β-D-glucopyranoside
Formula: C₁₅H₁₇NO₆
MW: 307.299 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063950
CAS Number: 29266-96-4