7-[4-(2,2-Diphosphonoethyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid dihydrobromide
Supplier: Apollo Scientific
Synonyms: Enoxacin bisphosphonate as dihydrobromide salt
BIA0220-1G BIA0220-5G
APOSBIA0220-1GEA 0 EUR
APOSBIA0220-1G APOSBIA0220-5G
7-[4-(2,2-Diphosphonoethyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid dihydrobromide
7-[4-(2,2-Diphosphonoethyl)piperazin-1-yl]-1-ethyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid dihydrobromide
Formula: C₁₇H₂₃FN₄O₉P₂·2BrH
MW: 670.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072177
CAS Number: 434341-03-4