15-Acetoxyscirpenol
Supplier: Apollo Scientific
Danger
Synonyms: 15-Acetoxyscirpendiol, 15-Acetoxyscirpen-3,4-diol
BIA2304-1MG
APOSBIA2304-1MGEA 133 EUR
APOSBIA2304-1MG
15-Acetoxyscirpenol
15-Acetoxyscirpenol
Formula: C₁₈H₂₆O₅
MW: 322.396 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Freezer
MDL Number: MFCD00078774
CAS Number: 2623-22-5
UN: 2811
ADR: 6.1,III