5-Amino-DL-tryptophan
Supplier: Apollo Scientific
Warning
BIA4013-250MG BIA4013-100MG BIA4013-500MG
APOSBIA4013-250MGEA 306 EUR
APOSBIA4013-250MG APOSBIA4013-100MG APOSBIA4013-500MG
5-Amino-DL-tryptophan
5-Amino-DL-tryptophan
Formula: C₁₁H₁₃N₃O₂
MW: 219.24 g/mol
Melting Pt: 270 °C
Storage Temperature: Freezer
MDL Number: MFCD02683918
CAS Number: 6383-69-3