Biotinyl-6-aminoquinoline
Supplier: Apollo Scientific
Warning
Synonyms: BAQ
BIB1154-10MG BIB1154-25MG BIB1154-5MG
APOSBIB1154-10MGEA 219 EUR
APOSBIB1154-10MG APOSBIB1154-25MG APOSBIB1154-5MG
Biotinyl-6-aminoquinoline
Biotinyl-6-aminoquinoline
Formula: C₁₉H₂₂N₄O₂S
MW: 370.469 g/mol
Melting Pt: 211…215 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063390
CAS Number: 91853-89-3