Bromophenol blue sodium salt
Supplier: Apollo Scientific
Warning
Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt
BIB1230-25G BIB1230-5G
APOSBIB1230-25GEA 72.5 EUR
APOSBIB1230-25G APOSBIB1230-5G
Bromophenol blue sodium salt
Bromophenol blue sodium salt
Formula: C₁₉H₉Br₄NaO₅S
MW: 691.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 62625-28-9