5-Bromo-4-chloro-3-indolyl nonanoate
Supplier: Apollo Scientific
Warning
Synonyms: X-nonanoate
BIB1403-250MG BIB1403-500MG
APOSBIB1403-250MGEA 267 EUR
APOSBIB1403-250MG APOSBIB1403-500MG
5-Bromo-4-chloro-3-indolyl nonanoate
5-Bromo-4-chloro-3-indolyl nonanoate
Formula: C₁₇H₂₁BrClNO₂
MW: 386.711 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02683900
CAS Number: 133950-77-3
UN: 3077
ADR: 9+EHS III