2-Naphthyl α-D-glucopyranoside

Supplier: Apollo Scientific
Warning

BIB1428-250MG BIB1428-500MG
APOSBIB1428-250MGEA 102 EUR
APOSBIB1428-250MG APOSBIB1428-500MG
2-Naphthyl α-D-glucopyranoside
2-Naphthyl α-D-glucopyranoside
Formula: C₁₆H₁₈O₆
MW: 306.311 g/mol
Melting Pt: 202 °C
Storage Temperature: Freezer
MDL Number: MFCD00067170
CAS Number: 25320-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR