L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt
Supplier: Apollo Scientific
Warning
Synonyms: L-Alanine-AMC.TFA
BIB1438-1G BIB1438-500MG
APOSBIB1438-1GEA 125 EUR
APOSBIB1438-1G APOSBIB1438-500MG
L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt
L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt
Formula: C₁₃H₁₄N₂O₃·CF₃CO₂H
MW: 360.285 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037394
CAS Number: 96594-10-4