Creatine phosphate disodium salt
Supplier: Apollo Scientific
Warning
Synonyms: Sodium creatine phosphate dibasic, N-Methyl-N-(N-phosphonocarbamimidoyl)glycine, disodium salt, Sodium creatine phosphate, Phosphocreatine disodium salt
BIC1452-25G BIC1452-5G
APOSBIC1452-25GEA 522 EUR
APOSBIC1452-25G APOSBIC1452-5G
Creatine phosphate disodium salt
Creatine phosphate disodium salt
Formula: C₄H₈N₃O₅P·2Na
MW: 255.077 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00014351
CAS Number: 6190-45-0