DST (Disuccinimidyl tartrate)

Supplier: Apollo Scientific
Warning

Synonyms: DST

APOSBICL108-1GEA 0 EUR
APOSBICL108-1G APOSBICL108-250MG
DST (Disuccinimidyl tartrate)
DST (Disuccinimidyl tartrate)

Primary amine reactive -Periodate cleavable


Ref: Farries,T.C.and Atkinson, J.P. (1989) J.Immun.142, 842-847Ref: Park,L.S,et al. (1986) J.Biol.Chem.261, 205-210

Formula: C₁₂H₁₂N₂O₁₀
MW: 344,231 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03788224
CAS Number: 77658-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR