1,6-Anhydro-2-azido-4-O-benzyl-2-deoxy-β-D-glucopyranose ≥99%
Supplier: Apollo Scientific
BICL4012-500MG BICL4012-100MG BICL4012-250MG
APOSBICL4012-500MGEA 864 EUR
APOSBICL4012-500MG APOSBICL4012-100MG APOSBICL4012-250MG
1,6-Anhydro-2-azido-4-O-benzyl-2-deoxy-β-D-glucopyranose ≥99%
1,6-Anhydro-2-azido-4-O-benzyl-2-deoxy-β-D-glucopyranose
Storage Temperature: Ambient
CAS Number: 55682-47-8