1,6-Anhydro-2-deoxy-2-fluoro-β-D-glucopyranose ≥99%
Supplier: Apollo Scientific
BICL4016-250MG BICL4016-500MG BICL4016-100MG
APOSBICL4016-250MGEA 696 EUR
APOSBICL4016-250MG APOSBICL4016-500MG APOSBICL4016-100MG
1,6-Anhydro-2-deoxy-2-fluoro-β-D-glucopyranose ≥99%
1,6-Anhydro-2-deoxy-2-fluoro-β-D-glucopyranose
Storage Temperature: Ambient
CAS Number: 23235-99-6