1,6-Anhydro-β-D-glucofuranose ≥98%
Supplier: Apollo Scientific
Warning
BICL4023-10MG BICL4023-5MG BICL4023-1MG
APOSBICL4023-10MGEA 1040 EUR
APOSBICL4023-10MG APOSBICL4023-5MG APOSBICL4023-1MG
1,6-Anhydro-β-D-glucofuranose ≥98%
1,6-Anhydro-β-D-glucofuranose
Formula: C₆H₁₀O₅
MW: 162.141 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Freezer
MDL Number: MFCD28014054
CAS Number: 7425-74-3