1,6-Anhydro-3,4-O-isopropylidene-β-D-galactopyranose ≥98%
Supplier: Apollo Scientific
BICL4030-1G BICL4030-2G BICL4030-500MG
APOSBICL4030-1GEA 538 EUR
APOSBICL4030-1G APOSBICL4030-2G APOSBICL4030-500MG
1,6-Anhydro-3,4-O-isopropylidene-β-D-galactopyranose ≥98%
1,6-Anhydro-3,4-O-isopropylidene-β-D-galactopyranose
Storage Temperature: Ambient
CAS Number: 52579-97-2