Diclofenac-acyl-β-D-glucuronide 95+%
Supplier: Apollo Scientific
Danger
BICL4062-1MG BICL4062-2MG
APOSBICL4062-1MGEA 490 EUR
APOSBICL4062-1MG APOSBICL4062-2MG
Diclofenac-acyl-β-D-glucuronide 95+%
Diclofenac-acyl-β-D-glucuronide
Melting Pt: 101…103 °C (dec.)
Storage Temperature: Freezer
CAS Number: 64118-81-6
UN: 2811
ADR: 6.1,III