1,2:5,6-Di-O-isopropylidene-3-O-tosyl-α-D-glucofuranose 99
Supplier: Apollo Scientific
Synonyms: NSC 14163, 1,2:5,6-Di-O-isopropylidene-3-O-toluenesulphonyl-α-D-glucofuranose
BICL4065-1G BICL4065-5G BICL4065-2G
APOSBICL4065-1GEA 420 EUR
APOSBICL4065-1G APOSBICL4065-5G APOSBICL4065-2G
1,2:5,6-Di-O-isopropylidene-3-O-tosyl-α-D-glucofuranose 99
1,2:5,6-Di-O-isopropylidene-3-O-tosyl-α-D-glucofuranose
Storage Temperature: Ambient
CAS Number: 3253-75-6