2,4-Di-O-acetyl-1,6-anhydro-3-O-benzyl-β-L-idopyranose ≥99%
Supplier: Apollo Scientific
BICL4078-250MG BICL4078-100MG
APOSBICL4078-250MGEA 906 EUR
APOSBICL4078-250MG APOSBICL4078-100MG
2,4-Di-O-acetyl-1,6-anhydro-3-O-benzyl-β-L-idopyranose ≥99%
2,4-Di-O-acetyl-1,6-anhydro-3-O-benzyl-β-L-idopyranose
Storage Temperature: Ambient
CAS Number: 61237-60-3