1,6:2,3-Dianhydro-4-O-(2',3',4',6'-tetra-O-acetyl-β-D-glucopyranosyl)-β-D-mannopyranose ≥99%
Supplier: Apollo Scientific
BICL4080-100MG BICL4080-250MG
APOSBICL4080-100MGEA 447 EUR
APOSBICL4080-100MG APOSBICL4080-250MG
1,6:2,3-Dianhydro-4-O-(2',3',4',6'-tetra-O-acetyl-β-D-glucopyranosyl)-β-D-mannopyranose ≥99%
1,6:2,3-Dianhydro-4-O-(2',3',4',6'-tetra-O-acetyl-β-D-glucopyranosyl)-β-D-mannopyranose
Storage Temperature: Ambient
CAS Number: 103702-87-0