Ethyl 2,3-di-O-benzyl-4,6-O-(R)-benzylidene-1-deoxy-1-thio-α-D-mannopyranoside S-oxide ≥98%
Supplier: Apollo Scientific
Synonyms: 2,3-Di-O-benzyl-4,6-O-(R)-benzylidene-1-deoxy-1-(ethylsulphinyl)-α-D-mannopyranoside
BICL4096-1G BICL4096-2G
APOSBICL4096-1GEA 504 EUR
APOSBICL4096-1G APOSBICL4096-2G
Ethyl 2,3-di-O-benzyl-4,6-O-(R)-benzylidene-1-deoxy-1-thio-α-D-mannopyranoside S-oxide ≥98%
Ethyl 2,3-di-O-benzyl-4,6-O-(R)-benzylidene-1-deoxy-1-thio-α-D-mannopyranoside S-oxide
Storage Temperature: Ambient
CAS Number: 188357-34-8