4-Methylphenyl 4,6-O-isopropylidene-1-thio-β-D-glucopyranoside ≥98%
Supplier: Apollo Scientific
Synonyms: Tolyl 4,6-O-isopropylidene-1-thio-β-D-glucopyranoside, Thiocresyl 4,6-O-isopropylidene-β-D-glucopyranoside
BICL4143-1G BICL4143-5G BICL4143-2G
APOSBICL4143-1GEA 238 EUR
APOSBICL4143-1G APOSBICL4143-5G APOSBICL4143-2G
4-Methylphenyl 4,6-O-isopropylidene-1-thio-β-D-glucopyranoside ≥98%
4-Methylphenyl 4,6-O-isopropylidene-1-thio-β-D-glucopyranoside
Storage Temperature: Ambient
CAS Number: 905716-02-1