4-Nitrophenyl 1-thio-β-D-glucopyranoside ≥98%
Supplier: Apollo Scientific
Warning
Synonyms: p-Nitrophenyl 1-thio-β-D-glucoside
BICL4181-100MG BICL4181-250MG BICL4181-50MG
APOSBICL4181-100MGEA 447 EUR
APOSBICL4181-100MG APOSBICL4181-250MG APOSBICL4181-50MG
4-Nitrophenyl 1-thio-β-D-glucopyranoside ≥98%
4-Nitrophenyl 1-thio-β-D-glucopyranoside
Melting Pt: 163…164 °C
Storage Temperature: Freezer
CAS Number: 2788-56-9