2,3,4-Tri-O-acetyl-1-amino-6-azido-1,6-dideoxy-β-D-glucopyranoside ≥98%
Supplier: Apollo Scientific
Synonyms: 2,3,4-Tri-O-acetyl-6-azido-1,6-dideoxy-β-D-glucopyranosylamine
BICL4214-50MG BICL4214-10MG BICL4214-25MG
APOSBICL4214-50MGEA 1010 EUR
APOSBICL4214-50MG APOSBICL4214-10MG APOSBICL4214-25MG
2,3,4-Tri-O-acetyl-1-amino-6-azido-1,6-dideoxy-β-D-glucopyranoside ≥98%
2,3,4-Tri-O-acetyl-1-amino-6-azido-1,6-dideoxy-β-D-glucopyranoside
Storage Temperature: Ambient
CAS Number: 565226-14-4