3,4,6-Tri-O-acetyl-1,2-dideoxy-1,2-difluoro-β-D-glucopyranose 99
Supplier: Apollo Scientific
Synonyms: 3,4,6-Tri-O-acetyl-1,2-dideoxy-2-fluoro-β-D-glucopyranosyl fluoride
BICL4216-100MG BICL4216-250MG BICL4216-500MG
APOSBICL4216-100MGEA 351 EUR
APOSBICL4216-100MG APOSBICL4216-250MG APOSBICL4216-500MG
3,4,6-Tri-O-acetyl-1,2-dideoxy-1,2-difluoro-β-D-glucopyranose 99
3,4,6-Tri-O-acetyl-1,2-dideoxy-1,2-difluoro-β-D-glucopyranose
Storage Temperature: Ambient
CAS Number: 29069-93-0