6-Azido-6-deoxy-2,3-O-isopropylidene-α-L-sorbofuranose ≥98%
Supplier: Apollo Scientific
BICL4222-1G BICL4222-2G BICL4222-5G
APOSBICL4222-1GEA 420 EUR
APOSBICL4222-1G APOSBICL4222-2G APOSBICL4222-5G
6-Azido-6-deoxy-2,3-O-isopropylidene-α-L-sorbofuranose ≥98%
6-Azido-6-deoxy-2,3-O-isopropylidene-α-L-sorbofuranose
Formula: C₉H₁₅N₃O₅
MW: 245.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144924
CAS Number: 126210-25-1