1,4-Anhydro-D-mannitol 98
Supplier: Apollo Scientific
BICL4260-100MG BICL4260-250MG
APOSBICL4260-100MGEA 447 EUR
APOSBICL4260-100MG APOSBICL4260-250MG
1,4-Anhydro-D-mannitol 98
1,4-Anhydro-D-mannitol
Formula: C₆H₁₂O₅
MW: 164.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702340
CAS Number: 7726-97-8