2,3,2',3',4',6'-Hexa-O-acetyl-1,6-anhydro-β-D-maltose ≥98%
Supplier: Apollo Scientific
Warning
Synonyms: NSC 105391, 1,6-Anhydro-2,3-di-O-acetyl-4-O-(2,3,4,6-tetra-O-acetyl-α-D-glucopyranosyl)-β-D-glucopyranose, Maltosan hexaacetate
BICL4297-1G BICL4297-250MG BICL4297-500MG
APOSBICL4297-1GEA 1200 EUR
APOSBICL4297-1G APOSBICL4297-250MG APOSBICL4297-500MG
2,3,2',3',4',6'-Hexa-O-acetyl-1,6-anhydro-β-D-maltose ≥98%
2,3,2',3',4',6'-Hexa-O-acetyl-1,6-anhydro-β-D-maltose
Formula: C₂₄H₃₂O₁₆
MW: 576.501 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28397787
CAS Number: 28868-67-9