2,3:4,5-Di-O-isopropylidene-D-arabinose ≥99%
Supplier: Apollo Scientific
Warning
Synonyms: D-Arabinose diacetonide
BICL4326-1G BICL4326-500MG BICL4326-2G
APOSBICL4326-1GEA 768 EUR
APOSBICL4326-1G APOSBICL4326-500MG APOSBICL4326-2G
2,3:4,5-Di-O-isopropylidene-D-arabinose ≥99%
2,3:4,5-Di-O-isopropylidene-D-arabinose
Formula: C₁₁H₁₈O₅
MW: 230.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144988
CAS Number: 13039-93-5