6-Deoxy-6-iodo-D-glucopyranose ≥98%
Supplier: Apollo Scientific
Synonyms: NSC 43143
BICL4331-1G BICL4331-500MG BICL4331-2G
APOSBICL4331-1GEA 1010 EUR
APOSBICL4331-1G APOSBICL4331-500MG APOSBICL4331-2G
6-Deoxy-6-iodo-D-glucopyranose ≥98%
6-Deoxy-6-iodo-D-glucopyranose
Formula: C₆H₁₁IO₅
MW: 290.053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28397796
CAS Number: 6304-86-5