6-O-Feruloyl-D-glucose ≥98%
Supplier: Apollo Scientific
Synonyms: 6-[(2E)-3-(4-Hydroxy-3-methoxyphenyl)-2-propenoate]-D-glucose
BICL4335-10MG BICL4335-25MG BICL4335-50MG
APOSBICL4335-10MGEA 336 EUR
APOSBICL4335-10MG APOSBICL4335-25MG APOSBICL4335-50MG
6-O-Feruloyl-D-glucose ≥98%
6-O-Feruloyl-D-glucose
Formula: C₁₆H₂₀O₉
MW: 356.325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18643048
CAS Number: 252207-50-4