4-Methylphenyl 4,6-O-benzylidene-1-thio-β-D-galactopyranoside ≥98%
Supplier: Apollo Scientific
|
Warning
|
BICL4356-1G
BICL4356-5G
BICL4356-2G
APOSBICL4356-1GEA
253
EUR
APOSBICL4356-1G
APOSBICL4356-5G
APOSBICL4356-2G
4-Methylphenyl 4,6-O-benzylidene-1-thio-β-D-galactopyranoside ≥98%
4-Methylphenyl 4,6-O-benzylidene-1-thio-β-D-galactopyranoside
Formula:
C₂₀H₂₂O₅S Melting Pt: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973630 CAS Number: 161007-96-1 |