1,2,3,4-Tetra-O-acetyl-6-O-trityl-β-D-glucopyranose ≥98%
Supplier: Apollo Scientific
Synonyms: NSC 404268, 1,2,3,4-Tetra-O-acetyl-6-O-triphenylmethyl-β-D-glucopyranose
BICL4370-1G BICL4370-500MG BICL4370-2G
APOSBICL4370-1GEA 490 EUR
APOSBICL4370-1G APOSBICL4370-500MG APOSBICL4370-2G
1,2,3,4-Tetra-O-acetyl-6-O-trityl-β-D-glucopyranose ≥98%
1,2,3,4-Tetra-O-acetyl-6-O-trityl-β-D-glucopyranose
Formula: C₃₃H₃₄O₁₀
MW: 590.617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076150
CAS Number: 37074-90-1