1,3,4,6-Tetra-O-acetyl-2-deoxy-2-iodo-D-galactopyranose ≥98%
Supplier: Apollo Scientific
BICL4372-100MG BICL4372-250MG BICL4372-500MG
APOSBICL4372-100MGEA 447 EUR
APOSBICL4372-100MG APOSBICL4372-250MG APOSBICL4372-500MG
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-iodo-D-galactopyranose ≥98%
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-iodo-D-galactopyranose
Formula: C₁₄H₁₉IO₉
MW: 458.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28010889
CAS Number: 1286737-83-4