Boc-L-homopropargylglycine
Supplier: Apollo Scientific
Warning
Synonyms: N,Boc-L-homopropargylglycine
BICR119-100MG BICR119-500MG
APOSBICR119-100MGEA 545 EUR
APOSBICR119-100MG APOSBICR119-500MG
Boc-L-homopropargylglycine
Boc-L-homopropargylglycine
Formula: C₁₁H₁₇NO₄
MW: 227.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052897
CAS Number: 208522-16-1