2C-Methyl-D-ribono-1,4-lactone
Supplier: Apollo Scientific
Warning
Synonyms: (3R,4R,5R)-3,4-Dihydroxy-5-(hydroxymethyl)-3-methyldihydrofuran-2(3H)-one
BIM1009-1G BIM1009-5G BIM1009-10G
APOSBIM1009-1GEA 21.9 EUR
APOSBIM1009-1G APOSBIM1009-5G APOSBIM1009-10G
2C-Methyl-D-ribono-1,4-lactone
2C-Methyl-D-ribono-1,4-lactone
Formula: C₆H₁₀O₅
MW: 162.141 g/mol
Melting Pt: 150…162 °C
Storage Temperature: Ambient
MDL Number: MFCD07369541
CAS Number: 492-30-8