Azido-PEG9-acid
Supplier: Apollo Scientific
BIPG1110-100MG BIPG1110-50MG BIPG1110-250MG
APOSBIPG1110-100MGEA 284 EUR
APOSBIPG1110-100MG APOSBIPG1110-50MG APOSBIPG1110-250MG
Azido-PEG9-acid
Azido-PEG9-acid
Formula: C₂₁H₄₁N₃O₁₁
MW: 511.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13184977
CAS Number: 1670249-37-2