Mal-amido-PEG12-NHS
Supplier: Apollo Scientific
Warning
BIPG1481-100MG BIPG1481-50MG
APOSBIPG1481-100MGEA 364 EUR
APOSBIPG1481-100MG APOSBIPG1481-50MG
Mal-amido-PEG12-NHS
Mal-amido-PEG12-NHS
Formula: C₃₈H₆₃N₃O₁₉
MW: 865.916 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11041148
CAS Number: 326003-46-7