m-PEG2-amine
Supplier: Apollo Scientific
Danger
Synonyms: 2-(2-Methoxyethoxy)ethanamine
BIPG1580-1G BIPG1580-5G
APOSBIPG1580-1GEA 121 EUR
APOSBIPG1580-1G APOSBIPG1580-5G
m-PEG2-amine
m-PEG2-amine
Formula: C₅H₁₃NO₂
Density: 0.958 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD09032941
CAS Number: 31576-51-9
UN: 2810
ADR: 6.1,III