t-Boc-N-Amido-PEG4-Amine
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl (14-amino-3,6,9,12-tetraoxatetradecyl)carbamate, Boc-NH-PEG4-CH2CH2NH2
BIPG1812-100MG BIPG1812-50MG BIPG1812-250MG
APOSBIPG1812-100MGEA 168 EUR
APOSBIPG1812-100MG APOSBIPG1812-50MG APOSBIPG1812-250MG
t-Boc-N-Amido-PEG4-Amine
t-Boc-N-Amido-PEG4-Amine
Formula: C₁₅H₃₂N₂O₆
MW: 336.424 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16619339
CAS Number: 811442-84-9