1-(2,2-Diethoxyethyl)piperazine 98%
Supplier: Apollo Scientific
Warning
Synonyms: (Piperazin-1-yl)acetaldehyde diethyl acetal
OR0080-1G OR0080-5G OR0080-25G
APOSOR0080-1GEA 71.2 EUR
APOSOR0080-1G APOSOR0080-5G APOSOR0080-25G
1-(2,2-Diethoxyethyl)piperazine 98%
1-(2,2-Diethoxyethyl)piperazine
Formula: C₁₀H₂₂N₂O₂
MW: 202.294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094589
CAS Number: 82516-06-1