4-(Dimethoxymethyl)-6-hydroxy-2-thiopyrimidine
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Dimethoxymethyl)-6-hydroxypyrimidine-2-thiol, 6-(Dimethoxymethyl)-2-sulphanylpyrimidin-4-ol
OR0091-1G OR0091-5G
APOSOR0091-1GEA 58 EUR
APOSOR0091-1G APOSOR0091-5G
4-(Dimethoxymethyl)-6-hydroxy-2-thiopyrimidine
4-(Dimethoxymethyl)-6-hydroxy-2-thiopyrimidine
Formula: C₇H₁₀N₂O₃S
MW: 202.231 g/mol
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00662704
CAS Number: 90370-38-0