3-(3,4-Dimethoxyphenyl)-1-(3-hydroxyphenyl)propan-1-one 95%
Supplier: Apollo Scientific
Warning
OR01535-500MG OR01535-250MG
APOSOR01535-500MGEA 249 EUR
APOSOR01535-500MG APOSOR01535-250MG
3-(3,4-Dimethoxyphenyl)-1-(3-hydroxyphenyl)propan-1-one 95%
3-(3,4-Dimethoxyphenyl)-1-(3-hydroxyphenyl)propan-1-one
Formula: C₁₇H₁₈O₄
MW: 286.322 g/mol
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD06245367
CAS Number: 178445-83-5