4-(3,5-Dimethyl-1H-pyrazol-1-yl)aniline
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Aminophenyl)-3,5-dimethyl-1H-pyrazole
OR01749-1G OR01749-5G
APOSOR01749-1GEA 98.6 EUR
APOSOR01749-1G APOSOR01749-5G
4-(3,5-Dimethyl-1H-pyrazol-1-yl)aniline
4-(3,5-Dimethyl-1H-pyrazol-1-yl)aniline
Formula: C₁₁H₁₃N₃
MW: 187.241 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00661746
CAS Number: 52708-32-4