2-Chloro-4,5-diamino-6-methylpyrimidine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-6-methylpyrimidine-4,5-diamine
OR01910-1G OR01910-5G
APOSOR01910-1GEA 114 EUR
APOSOR01910-1G APOSOR01910-5G
2-Chloro-4,5-diamino-6-methylpyrimidine 95%
2-Chloro-4,5-diamino-6-methylpyrimidine
Formula: C₅H₇ClN₄
Melting Pt: 249…252 °C
Storage Temperature: Ambient
MDL Number: MFCD01764685
CAS Number: 63211-98-3
UN: 2811
ADR: 6.1,III