1,3-Diazaspiro[4.5]decane-2,4-dione
Supplier: Apollo Scientific
Warning
OR0331-1G OR0331-250MG OR0331-5G
APOSOR0331-1GEA 59.6 EUR
APOSOR0331-1G APOSOR0331-250MG APOSOR0331-5G
1,3-Diazaspiro[4.5]decane-2,4-dione
1,3-Diazaspiro[4.5]decane-2,4-dione
Formula: C₈H₁₂N₂O₂
MW: 168.193 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00022404
CAS Number: 702-62-5