[4-(Thien-2-yl)phenyl]methanol
Supplier: Apollo Scientific
Warning
Synonyms: 2-[4-(Hydroxymethyl)phenyl]thiophene, 4-(Thien-2-yl)benzyl alcohol, (4-(Thiophen-2-yl)phenyl)methanol
OR0464-1G OR0464-250MG
APOSOR0464-1GEA 126 EUR
APOSOR0464-1G APOSOR0464-250MG
[4-(Thien-2-yl)phenyl]methanol
[4-(Thien-2-yl)phenyl]methanol
Formula: C₁₁H₁₀OS
Boiling Pt: 311.5 °C
Melting Pt: 60…61 °C
Flash Pt: 115.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06203037
CAS Number: 81443-44-9