2-Methyl-1-[3-(morpholin-4-yl)prop-1-yl]-5-phenyl-1H-pyrrole-3-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 3-Carboxy-2-methyl-1-[3-(morpholin-4-yl)prop-1-yl]-5-phenyl-1H-pyrrole, 4-[3-(3-Carboxy-2-methyl-5-phenyl-1H-pyrrol-1-yl)prop-1-yl]morpholine
OR0634-1G OR0634-5G
APOSOR0634-1GEA 229 EUR
APOSOR0634-1G APOSOR0634-5G
2-Methyl-1-[3-(morpholin-4-yl)prop-1-yl]-5-phenyl-1H-pyrrole-3-carboxylic acid
2-Methyl-1-[3-(morpholin-4-yl)prop-1-yl]-5-phenyl-1H-pyrrole-3-carboxylic acid
Formula: C₁₉H₂₄N₂O₃
MW: 328.405 g/mol
Melting Pt: 169…177 °C
Storage Temperature: Ambient
MDL Number: MFCD01567236
CAS Number: 306936-20-9